Order Entry
Finland
Orders LinkContactUsLinkComponent
3,4-Dichlorophenol ≥99%
3,4-Dichlorophenol ≥99%
Glass bottle for solids, 25 g
  A12256.14
3,4-Dichlorophenol ≥99%
  A12256.14
 :  95-77-2
3,4-Dichlorophenol ≥99%
3,4-Dichlorophenol ≥99%
Glass bottle for solids, 25 g
 :  95-77-2
162,00 €
1 * 25 g
Purity: 99% CAS No.: 95-77-2 Molecular Formula: C6H4Cl2O Formula Weight: 163.00 Odour: Phenol-like Melting Point: 64-68° Boiling Point: 252-255° Beilstein Reference: 1907693 UN No.: 2020 MDL No.: MFCD00002258
  •  :95-77-2
  •  :C₆H₄Cl₂O
  •  :MFCD00002258
  •  :163 g/mol
  •  :252…255 °C
  •  :64…68 °C
  •  :1.6 g/cm³ (20 °C)
  •  :Ambient
  •  :202-450-5
  •  :2020
  •  :6.1,III
  •  :Density: 1.6 g/cm³ (20 °C)
  • Pack type:
    Glass bottle for solids
  • Pk:
    25 g