Order Entry
Finland
Orders LinkContactUsLinkComponent
Ethanolamine ≥98%

Ethanolamine ≥98%

Glass bottle for solids, 500 g
Catalog # A11697.36
Corrosive
Irritant
Ethanolamine ≥98%
Catalog # A11697.36
CAS Number:  141-43-5
Ethanolamine ≥98%
Ethanolamine ≥98%

Glass bottle for solids, 500 g

CAS Number:  141-43-5
Price
66,40 €
Log in for availability
1 * 500 g
Product Details & Documents

Purity: 98+% CAS No.: 141-43-5 Molecular Formula: C2H7NO Formula Weight: 61.08 Flash Point: 93°(199°F) Refractive Index: 1.4540 Odour: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:14,3727
  • Boiling Pt:169…170 °C
  • Melting Pt:10 °C
  • Density:1.012
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.