Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Ethanolamine ≥98%

Glass bottle for solids, 2.500 g
Catalog # A11697.0E
Corrosive
Irritant
Ethanolamine ≥98%
Catalog # A11697.0E
CAS Number:  141-43-5
Ethanolamine ≥98%
Ethanolamine ≥98%

Glass bottle for solids, 2.500 g

CAS Number:  141-43-5
Price
155,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 98+% CAS No.: 141-43-5 Molecular Formula: C2H7NO Formula Weight: 61.08 Flash Point: 93°(199°F) Refractive Index: 1.4540 Odour: Ammonia-like Melting Point: 10° Boiling Point: 169-170° Beilstein Reference: 505944 Merck Reference: 14,3727 UN No.: 2491 MDL No.: MFCD00008183

Synonyms:  2-Aminoethanol, Monoethanolamine
  • CAS Number:141-43-5
  • Formula:NH₂CH₂CH₂OH
  • MDL Number:MFCD00008183
  • Molecular Weight:61.08 g/mol
  • Merck Index:14,3727
  • Boiling Pt:169…170 °C
  • Melting Pt:10 °C
  • Density:1.012
  • Flash Pt:93 °C (199 °F)
  • Storage Temperature:Ambient
  • EINECS:205-483-3
  • UN:2491
  • ADR:8,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications