Order Entry
Finland
Orders LinkContactUsLinkComponent
Benzophenone ≥99%

Benzophenone ≥99%

Glass bottle for solids, 250 g
Catalog # A10739.30
Environment
Benzophenone ≥99%
Catalog # A10739.30
CAS Number:  119-61-9
Benzophenone ≥99%
Benzophenone ≥99%

Glass bottle for solids, 250 g

CAS Number:  119-61-9
Price
54,80 €
Log in for availability
1 * 250 g
Product Details & Documents

Purity: 99% CAS No.: 119-61-9 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 138°(280°F) Physical Form: Cryst flakes or powder Density: 1.111 Appearance: White Odour: Pleasant Melting Point: 47-50° Boiling Point: 305° Beilstein Reference: 1238185 Merck Reference: 14,1098 UN No.: 3077 MDL No.: MFCD00003076

Synonyms:  Diphenylmethanone, Diphenyl ketone
  • CAS Number:119-61-9
  • Formula:C₁₃H₁₀O
  • MDL Number:MFCD00003076
  • Molecular Weight:182.22 g/mol
  • Merck Index:14,1098
  • Boiling Pt:305 °C
  • Melting Pt:47…50 °C
  • Density:1.111
  • Flash Pt:138 °C (280 °F)
  • Storage Temperature:Ambient
  • EINECS:204-337-6
  • UN:3077
  • ADR:9,III
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.