Benzophenone ≥99%
Glass bottle for solids, 1.000 g
Supplier: Thermo Fisher Scientific
Jump to:
Product Details & Documents
Purity: 99% CAS No.: 119-61-9 Molecular Formula: C13H10O Formula Weight: 182.22 Flash Point: 138°(280°F) Physical Form: Cryst flakes or powder Density: 1.111 Appearance: White Odour: Pleasant Melting Point: 47-50° Boiling Point: 305° Beilstein Reference: 1238185 Merck Reference: 14,1098 UN No.: 3077 MDL No.: MFCD00003076
Synonyms: Diphenylmethanone, Diphenyl ketone
- CAS Number:119-61-9
- Formula:C₁₃H₁₀O
- MDL Number:MFCD00003076
- Molecular Weight:182.22 g/mol
- Merck Index:14,1098
- Boiling Pt:305 °C
- Melting Pt:47…50 °C
- Density:1.111
- Flash Pt:138 °C (280 °F)
- Storage Temperature:Ambient
- EINECS:204-337-6
- UN:3077
- ADR:9,III
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1.000 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



