Order Entry
Finland
Orders LinkContactUsLinkComponent
Propionic acid ≥99%

Propionic acid ≥99%

Glass bottle for solids, 2.500 g
Catalog # L04210.0E
Corrosive
Flammable
Propionic acid ≥99%
Catalog # L04210.0E
CAS Number:  79-09-4
Propionic acid ≥99%
Propionic acid ≥99%

Glass bottle for solids, 2.500 g

CAS Number:  79-09-4
Price
125,00 €
Log in for availability
1 * 2.500 g
Product Details & Documents

Purity: 99% CAS No.: 79-09-4 Molecular Formula: C3H6O2 Formula Weight: 74.08 Flash Point: 51°(123°F) Refractive Index: 1.3875 Odour: Unpleasant Melting Point: -21° Boiling Point: 140-141° Beilstein Reference: 506071 Merck Reference: 14,7825 UN No.: 3463 MDL No.: MFCD00002756

Synonyms:  Propanoic acid
  • CAS Number:79-09-4
  • Formula:C₃H₆O₂
  • MDL Number:MFCD00002756
  • Molecular Weight:74.08 g/mol
  • Merck Index:14,7825
  • Boiling Pt:140…141 °C
  • Melting Pt:–21 °C
  • Density:0.992
  • Flash Pt:51 °C (123 °F)
  • Storage Temperature:Ambient
  • EINECS:201-176-3
  • UN:3463
  • ADR:8,II
  • REACH:01-2119486971-24
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    2.500 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.