Order Entry
Finland
ContactUsLinkComponent
Universal-reagenz reservoir, pp, reservoir is used on both sides. on the one hand, with reservoir 50 ml, on the other pages with a s plit 8-or 12-reservoir with capacity 5 per division 1 * 1 items
Tuotenumero SEMA5573
Supplier:  SEMADENI
Universal-reagenz reservoir, pp, reservoir is used on both sides. on the one hand, with reservoir 50 ml, on the other pages with a s plit 8-or 12-reservoir with capacity 5 per division 1 * 1 items
Tuotenumero SEMA5573
Supplier:  SEMADENI
Frequently Bought Together