Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
GRADE 17CHR CIRCLES, 70MM 100/PK 1 * 100 items
Catalog # WHAT1017-070
Supplier:
Whatman products (Cytiva)
Expand image
GRADE 17CHR CIRCLES, 70MM 100/PK 1 * 100 items
Catalog # WHAT1017-070
Supplier:
Whatman products (Cytiva)