Order Entry
Finland
Orders LinkContactUsLinkComponent
Caps with dropper inserts for bottles
Bottles with GL 18 thread
Catalog # VITL83306
Supplier:   VITLAB
Caps with dropper inserts for bottles
Catalog # VITL83306
Supplier:  VITLAB
Caps with dropper inserts for bottles
Caps with dropper inserts for bottles
Bottles with GL 18 thread
Price
97,00 €
Log in for availability
1 * 24 Items
Product Details & Documents
Made from HDPE.
  • Extra long, fine dropping tip for accurate dispensing
  • Good chemical resistance
  • Good temperature resistance
For bottles with GL threads. Cap with dropper insert, complete with screw cap.
Specifications
  • For:
    Bottles with GL 18 thread
  • Seq:
    01