Order Entry
Finland
Orders LinkContactUsLinkComponent
Watch dishes
Watch dishes
Watch glass, PTFE, without base, 125 mm
Catalog # VITL113497
Supplier:   VITLAB
Watch dishes
Catalog # VITL113497
Supplier:  VITLAB
Watch dishes
Watch dishes
Watch glass, PTFE, without base, 125 mm
Price
78,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
Watch dishes, made of PTFE, white.
  • Without base
  • Very good chemical resistance
  • Good temperature resistance
Ideal for use to cover beakers.
Certifications: DIN EN 285
Specifications
  • Ø:
    125 mm
  • Description:
    Watch glass, PTFE, without base
  • Seq:
    04