Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
TUBE WASSERSTANDANZ. 9,5 MM - 100 PC 1 * 16,8 kg
Catalog # SCOR1026278
Supplier:
Schott AG
Expand image
TUBE WASSERSTANDANZ. 9,5 MM - 100 PC 1 * 16,8 kg
Catalog # SCOR1026278
Supplier:
Schott AG