Sumatriptan Related Compounds, Reference Standard
Sumatriptan Succinate Related Compound A, USP reference standard, Vial, 15 mg
Tuotenumero 1642212.
Toimittaja: USP
Siirry kohtaan:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
Certifications: US Pharmacopea Reference Standard
Tekniset tiedot
Tekniset tiedot
- Pack type:Vial
- Pk:15 mg
- Standards:USP reference standard
- Description:Sumatriptan Succinate Related Compound A
- Cat. No.:1642212.
- Formula:C₂₇H₃₇N₅O₂S
- Description:Sumatriptan Succinate Related Compound A
- Synonym(s):[3-[2-(dimethylamino)ethyl]-2-[[3-[2-(dimethylamino)ethyl]-1H-indol-5-yl]methyl]-1H-indol-5-yl]-N-methylmethanesulfonamide, succinate salt
- CAS No.:545338-89-4
Tekniset tiedot
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



