Hydroxyzine Related Compounds, Reference Standard
Hydroxyzine Related Compound A, USP reference standard, Vial, 25 mg
Supplier: USP
Jump to:
Product Details & Documents
USP Reference Standards are specified for use in conducting official USP–NF tests and assays. USP also provides Reference Standards specified in the Food Chemicals Codex as well as authentic substances—high-quality chemical samples—as a service to analytical, clinical, pharmaceutical and research laboratories. To confirm accuracy and reproducibility, USP Reference Standards are rigorously tested and evaluated by multiple independent laboratories including USP, commercial, regulatory, and academic labs. USP also provide publicly available, official documentary standards for pharmaceutical ingredients in the USP–NF that link directly with our primary reference standards.
: US Pharmacopea Reference Standard
Certificates
Search by Lot NumberSpecifications
Specifications
- Pack type:Vial
- Pk:25 mg
- Standards:USP reference standard
- Description:Hydroxyzine Related Compound A
- Cat. No.:1333058.
- Formula:C₁₇H₁₉ClN₂
- Description:Hydroxyzine Related Compound A
- Synonym(s):1-[(4-Chlorophenyl)phenylmethyl]piperazine
- CAS No.:303-26-4



