Order Entry
Finland
Orders LinkContactUsLinkComponent
Assortment Boxes
Assortment Boxes
Box, A5 size, 60 mm high, no lid, 224×175×60 mm
Catalog # 216-0674
Supplier:   HOFSTATTER EBBESEN
Assortment Boxes
Catalog # 216-0674
Supplier:  HOFSTATTER EBBESEN
Assortment Boxes
Assortment Boxes
Box, A5 size, 60 mm high, no lid, 224×175×60 mm
Price
34,00 €
Log in for availability
1 * 1 Items
Product Details & Documents
PS, clear.
Boxes in various sizes and heights. Ideal for storing small parts.
  • Compartment partitions for flexible compartment sizes
  • Clearly arranged storage for small materials
  • Lightweight and easy to clean
Specifications
  • Description:
    Box, A5 size, 60 mm high, no lid
  • W×D×H:
    224×175×60 mm
Accessories
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, long, no slots, for A4 size, 60 mm high
    Partition, long, no slots, for A4 size, 60 mm high
    216-0663
    56 mm
    304 mm
    Log in for availability
    1 * 1 Items - 6,80 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 11 slots, for A4 size, 60 mm high
    Partition with 11 slots, for A4 size, 60 mm high
    216-0664
    56 mm
    304 mm
    Log in for availability
    1 * 1 Items - 7,50 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, long, no slots, for A4 size, 60 mm high
    Partition, long, no slots, for A4 size, 60 mm high
    216-0672
    56 mm
    304 mm
    Log in for availability
    1 * 1 Items - 11,00 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, short, no slots, for A4 size, 60 mm high
    Partition, short, no slots, for A4 size, 60 mm high
    216-0667
    56 mm
    108 mm
    Log in for availability
    1 * 1 Items - 3,00 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, short, no slots, for A6 size, 40 mm high
    Partition, short, no slots, for A6 size, 40 mm high
    216-0668
    34 mm
    53 mm
    Log in for availability
    1 * 1 Items - 8,00 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, short, no slots, for A4 size, 60 mm high
    Partition, short, no slots, for A4 size, 60 mm high
    216-0677
    56 mm
    71 mm
    Log in for availability
    1 * 1 Items - 5,20 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, short, no slots, for A6 size, 40 mm high
    Partition, short, no slots, for A6 size, 40 mm high
    216-0678
    34 mm
    34 mm
    Log in for availability
    1 * 1 Items - 4,60 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, short, no slots, for A4 size, 60 mm high
    Partition, short, no slots, for A4 size, 60 mm high
    216-0679
    56 mm
    53 mm
    Log in for availability
    1 * 1 Items - 3,40 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Lid, PS, clear, for A4 size
    Lid, PS, clear, for A4 size
    216-0680
    12 mm
    315 mm
    Log in for availability
    1 * 1 Items - 39,80 €
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Lid, PS, clear, for A5 size
    Lid, PS, clear, for A5 size
    216-0681
    178 mm
    12 mm
    Log in for availability
    1 * 1 Items - 20,40 €
Product Information
  • Catalog #
    Width
    Height
    Availability
    Price
  • Lid, PS, clear, for A6 size
    Lid, PS, clear, for A6 size
    216-0682
    117 mm
    12 mm
    Log in for availability
    1 * 1 Items - 13,80 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, no slots, for A6 size, 60 mm high
    Partition, no slots, for A6 size, 60 mm high
    216-0650
    56 mm
    106 mm
    Log in for availability
    1 * 1 Items - 5,20 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 3 slots, for A6 size, 60 mm high
    Partition with 3 slots, for A6 size, 60 mm high
    216-0651
    56 mm
    106 mm
    Log in for availability
    1 * 1 Items - 5,10 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, long, no slots, for A6 size, 40 mm high
    Partition, long, no slots, for A6 size, 40 mm high
    216-0652
    34 mm
    107 mm
    Log in for availability
    1 * 1 Items - 2,60 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 5 slots, for A5 and A6 size, 60 mm high
    Partition with 5 slots, for A5 and A6 size, 60 mm high
    216-0654
    56 mm
    169 mm
    Log in for availability
    1 * 1 Items - 4,80 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 5 slots, for A5 and A6 size, 40 mm high
    Partition with 5 slots, for A5 and A6 size, 40 mm high
    216-0655
    41 mm
    170 mm
    Log in for availability
    1 * 1 Items - 4,20 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 5 slots, for A4 size, 120 mm high
    Partition with 5 slots, for A4 size, 120 mm high
    216-0656
    124 mm
    215 mm
    Log in for availability
    1 * 1 Items - 14,70 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition, medium, no slots, for A4 and A5 size, 60 mm high
    Partition, medium, no slots, for A4 and A5 size, 60 mm high
    216-0657
    56 mm
    218 mm
    Log in for availability
    1 * 1 Items - 5,50 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 7 slots, for A5 size, 40 mm high
    Partition with 7 slots, for A5 size, 40 mm high
    216-0659
    41 mm
    219 mm
    Log in for availability
    1 * 1 Items - 7,20 €
Product Information
  • Catalog #
    Height
    Length
    Availability
    Price
  • Partition with 5 slots, for A4 and A5 size, 60 mm high
    Partition with 5 slots, for A4 and A5 size, 60 mm high
    216-0658
    56 mm
    218 mm
    Log in for availability
    1 * 1 Items - 5,80 €