Order Entry
Finland
Orders LinkContactUsLinkComponent
Roller coaster
Roller coaster
Trolley to the runway
Catalog # 765-1307
Marketsource
Supplier:   PHYWE
Roller coaster
Catalog # 765-1307
Supplier:  PHYWE
Marketsource
Roller coaster
Roller coaster
Trolley to the runway
Price
694,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
The profile of the runway has been designed so that the experiments can be performed easily, quickly and safely. The length of the track (1,5 m) is more than sufficient for all conceivable physical experiments to be performed. The trolleys are equipped with overload protection in the form of a recessed, resilient wheel suspension. Characteristics.
  • Quick-lock system means easy and quick installation of
  • the light ports on the track without screws
  • PHYWES standard light ports are used which can also be used for stand-alone experiments
  • The footrests can be adjusted along the entire course, which enables trouble-free and safe installation on small surfaces
  • The track is easily adjusted in balance with the footrests' adjusting feet
  • The measuring scale is integrated in the track
  • There is a wide range of accessories for the runway
Specifications
  • Description:
    Trolley to the runway