Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Lecithin

Glass bottle for solids, 250 g
Catalog # ROTH9812.1
Marketsource
Supplier:   Roth Carl
Lecithin
Catalog # ROTH9812.1
Supplier:  Roth Carl
Marketsource
CAS Number:  8002-43-5
Lecithin

Glass bottle for solids, 250 g

CAS Number:  8002-43-5
Price
61,29 €
Log in for availability
1 * 250 g
Product Details & Documents
  • CAS Number:8002-43-5
  • MDL Number:MFCD00147406
  • Merck Index:12,05452
  • Density:1.0305 g/cm³ (20 °C)
  • EINECS:232-307-2

 

Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    250 g

Specifications