Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD C32 1 * 470 ml
Catalog # CANN9727-C32_U
Supplier:
CANNON INSTRUMENT
Expand image
VISCOSITY STANDARD C32 1 * 470 ml
Catalog # CANN9727-C32_U
Supplier:
CANNON INSTRUMENT