Order Entry
Finland
Orders LinkContactUsLinkComponent
Pendulum bobs
Pendulum bobs
Steel pendulum bob, 12,7 mm
Catalog # 765-1154
Marketsource
Supplier:   PHYWE
Pendulum bobs
Catalog # 765-1154
Supplier:  PHYWE
Marketsource
Pendulum bobs
Pendulum bobs
Steel pendulum bob, 12,7 mm
Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents
Suitable for use as pendulum bobs for frequency experiments. Hardened and polished ball bearings with eyelet.
Specifications
  • Description:
    Steel pendulum bob
  • Ø:
    12,7 mm