Order Entry
Finland
Orders LinkContactUsLinkComponent
Conductor balls

Conductor balls

Conductor ball, diameter 120 mm
Catalog # 765-1192
Marketsource
Supplier:   PHYWE
Conductor balls
Catalog # 765-1192
Supplier:  PHYWE
Marketsource
Conductor balls
Conductor balls

Conductor ball, diameter 120 mm

Price
484,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

For electrostatic experiments, for example, determination of the charge of a sphere and induction experiments. Hollow surface-treated brass sphere equipped with 4 mm holes for connecting banana connectors.

Specifications
  • Description:
    Conductor ball, diameter 120 mm