Order Entry
Finland
Orders LinkContactUsLinkComponent
Conductor balls

Conductor balls

Conductor ball, diameter 40 mm
Catalog # 765-1191
Marketsource
Supplier:   PHYWE
Conductor balls
Catalog # 765-1191
Supplier:  PHYWE
Marketsource
Conductor balls
Conductor balls

Conductor ball, diameter 40 mm

Price
160,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

For electrostatic experiments, for example, determination of the charge of a sphere and induction experiments. Hollow surface-treated brass sphere equipped with 4 mm holes for connecting banana connectors.

Specifications
  • Description:
    Conductor ball, diameter 40 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.