Molecular models - scale atoms
Scale atom I
764-0063
Marketsource
Scale atoms I , laboratory kit with 69 scale atoms and 16 lids in a box. Scale atoms II , laboratory kit with 109 scale atoms and 24 lids in a box. Scale atoms III >, laboratory kit with 293 scale atoms, 34 lids and 100 locking pins in a box.
- Overview of details in the scale turnover rates
- Description:Scale atom I