Order Entry
Finland
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Tray carrier with handle, —, - mm
Catalog # 765-0964
Supplier:   GRATNELL'S
Storage Boxes
Catalog # 765-0964
Supplier:  GRATNELL'S
Storage Boxes
Storage Boxes

Tray carrier with handle, —, - mm

Price
Inquire For Price
Log in for availability
1 * 1 Items
Product Details & Documents

Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.

  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable
Specifications
  • Colour:
  • Description:
    Tray carrier with handle
  • W×D×H:
    - mm