Order Entry
Finland
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Trays, Green, 312×430×300 mm
Catalog # 763-0309
Supplier:   GRATNELL'S
Storage Boxes
Catalog # 763-0309
Supplier:  GRATNELL'S
Storage Boxes
Storage Boxes

Trays, Green, 312×430×300 mm

Price
50,20 €
Log in for availability
1 * 1 Items
Product Details & Documents

Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.

  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable
Specifications
  • Colour:
    Green
  • Description:
    Trays
  • W×D×H:
    312×430×300 mm
Suositukset

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.