Order Entry
Finland
Orders LinkContactUsLinkComponent
Storage Boxes

Storage Boxes

Trays, Green, 312×427×150 mm
Catalog # 763-0305
Supplier:   GRATNELL'S
Storage Boxes
Catalog # 763-0305
Supplier:  GRATNELL'S
Storage Boxes
Storage Boxes

Trays, Green, 312×427×150 mm

Price
35,00 €
Log in for availability
1 * 1 Items
Product Details & Documents

Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.

  • BSI tested and passed for heavy educational use
  • All trays fit standard school furniture
  • Lids and inserts also available
  • Fully recyclable
Specifications
  • Colour:
    Green
  • Description:
    Trays
  • W×D×H:
    312×427×150 mm