Order Entry
Finland
Orders LinkContactUsLinkComponent
Forceps, pointed
Forceps, pointed
Straight, 105 mm
Catalog # 232-0125
Supplier:   COMACO HANDEL
Forceps, pointed
Catalog # 232-0125
Supplier:  COMACO HANDEL
Forceps, pointed
Forceps, pointed
Straight, 105 mm
Price
9,80 €
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel, grade 18-8.
  • Pointed
  • With serrated grip and tips
Specifications
  • Type:
    Straight
  • Length:
    105 mm
  • Seq:
    01