Order Entry
Finland
Orders LinkContactUsLinkComponent
Forceps, blunt
Forceps, blunt
Tweezers, 115 mm
Catalog # 232-0119
Supplier:   COMACO HANDEL
Forceps, blunt
Catalog # 232-0119
Supplier:  COMACO HANDEL
Forceps, blunt
Forceps, blunt
Tweezers, 115 mm
Price
9,50 €
Log in for availability
1 * 1 Items
Product Details & Documents
Stainless steel, grade 18-8.
  • Blunt tips, grip and tips serrated
Specifications
  • Description:
    Tweezers
  • Length:
    115 mm