Order Entry
Finland
Orders LinkContactUsLinkComponent
Hexadecan-16-olide ≥97%

Hexadecan-16-olide ≥97%

Glass bottle for solids, 50 g
Catalog # L03864.18
Irritant
Hexadecan-16-olide ≥97%
Catalog # L03864.18
CAS Number:  109-29-5
Hexadecan-16-olide ≥97%
Hexadecan-16-olide ≥97%

Glass bottle for solids, 50 g

CAS Number:  109-29-5
Price
602,00 €
Log in for availability
1 * 50 g
Product Details & Documents

Purity: 97% CAS No.: 109-29-5 Molecular Formula: C16H30O2 Formula Weight: 254.42 Flash Point: 100°(212°F) Melting Point: 33-34° Beilstein Reference: 148631 MDL No.: MFCD00039668

Synonyms:  Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide
  • CAS Number:109-29-5
  • Formula:C₁₆H₃₀O₂
  • MDL Number:MFCD00039668
  • Molecular Weight:254.41 g/mol
  • Melting Pt:33…34 °C
  • Flash Pt:>100 °C (212 °F)
  • Storage Temperature:Ambient
  • EINECS:203-662-0
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    50 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.