Order Entry
Finland
Orders LinkContactUsLinkComponent
Hexadecan-16-olide ≥97%
Glass bottle for solids, 10 g
  L03864.09
Hexadecan-16-olide ≥97%
  L03864.09
 :  109-29-5
Hexadecan-16-olide ≥97%
Hexadecan-16-olide ≥97%
Glass bottle for solids, 10 g
 :  109-29-5
182,00 €
1 * 10 g
Purity: 97% CAS No.: 109-29-5 Molecular Formula: C16H30O2 Formula Weight: 254.42 Flash Point: 100°(212°F) Melting Point: 33-34° Beilstein Reference: 148631 MDL No.: MFCD00039668
 :  Oxa-2-cycloheptadecanone, Hexadecanolactone, Oxacycloheptadecan-2-one, 16-Hexadecanolide
  •  :109-29-5
  •  :C₁₆H₃₀O₂
  •  :MFCD00039668
  •  :254.41 g/mol
  •  :33…34 °C
  •  :>100 °C (212 °F)
  •  :Ambient
  •  :203-662-0
  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g