Order Entry
Finland
Orders LinkContactUsLinkComponent
Readycult®

Readycult®

Readycult® Enterococci 100**, 20 Tests
Catalog # 1.01299.0001
Supplier:   Merck
Readycult®
Catalog # 1.01299.0001
Supplier:  Merck
CAS Number:  
Readycult®
Readycult®

Readycult® Enterococci 100**, 20 Tests

CAS Number:  
Price
175,00 €
Log in for availability
1 * 1 Each
Product Details & Documents

Readycult® Coliform and Readycult® Enterococci make it easy to detect coliforms, E. coli or faecal streptococci in water. You only need a sterile container and a UV lamp (Cat. No. 1.13203.0001).

  • Easy handling: Readycult® is ready to use in a blister pack
  • Quick results: Add Readycult® to the water sample straight from the blister pack, mix, incubate for 24 ±1 hour, read result, finished!
  • Safe and easy interpretation: Readycult® reliably detects even damaged organisms. Blue-green colour change shows the presence of Total coliforms, E. coli or Enterococci (Cat. No. 1.01299); fluorescence + indole reaction shows the presence of E. coli. * for 50 ml water; ** for 100 ml water
Specifications

Specifications

  • Pk:
    20 Tests
  • Description:
    Readycult® Enterococci 100**

Specifications