3-(2-Hydroxyethyl)indole ≥97%
Glass bottle for solids, 5 g
Supplier: Thermo Fisher Scientific
3-(2-Hydroxyethyl)indole ≥97%
Catalog # L02555.06
Supplier: Thermo Fisher Scientific
CAS Number: 526-55-6
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659
Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
- CAS Number:526-55-6
- Formula:C₁₀H₁₁NO
- MDL Number:MFCD00005659
- Molecular Weight:161.2 g/mol
- Merck Index:14,9798
- Boiling Pt:174 °C (2 mmHg)
- Melting Pt:56…59 °C
- Flash Pt:255 °C (491 °F)
- Storage Temperature:Ambient
- EINECS:208-393-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:5 g



