3-(2-Hydroxyethyl)indole ≥97%
Glass bottle for solids, 1 g
Toimittaja: Thermo Fisher Scientific
3-(2-Hydroxyethyl)indole ≥97%
Tuotenumero L02555.03
Toimittaja: Thermo Fisher Scientific
CAS numero: 526-55-6
Jump to:
Product Details & Documents
Purity: 97% CAS No.: 526-55-6 Molecular Formula: C10H11NO Formula Weight: 161.20 Flash Point: 255°(491°F) Melting Point: 56-59° Boiling Point: 174°/2mm Beilstein Reference: 125553 Merck Reference: 14,9798 MDL No.: MFCD00005659
Synonyms: 3-Indoleethanol, Indol-3-ethanol, Tryptophol, β-Indol-3-ylethanol
- CAS Number:526-55-6
- Formula:C₁₀H₁₁NO
- MDL Number:MFCD00005659
- Molecular Weight:161.2 g/mol
- Merck Index:14,9798
- Boiling Pt:174 °C (2 mmHg)
- Melting Pt:56…59 °C
- Flash Pt:255 °C (491 °F)
- Storage Temperature:Ambient
- EINECS:208-393-2
Documents
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:1 g
Specifications
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



