Storage Boxes
Trays, Translucent, 312×430×300 mm
Catalog # 763-0314
Supplier: GRATNELL'S
Jump to:
Product Details & Documents
Polypropylene storage trays. The material is fire-resistant and resistant to most chemicals in the school environment. Stackable.
- BSI tested and passed for heavy educational use
- All trays fit standard school furniture
- Lids and inserts also available
- Fully recyclable
Specifications
- Colour:Translucent
- Description:Trays
- W×D×H:312×430×300 mm



