Order Entry
Finland
ContactUsLinkComponent
Container, straight, capacity 180ml, material natural polystyrene, diameter 52mm, height 102mm, color of screw cap red, shapolyethyl ene assembled, CE, IVD, clinical and industrial, sterile. 1 * 264 items
Catalog # 216-2601
Supplier:  VWR Collection
Container, straight, capacity 180ml, material natural polystyrene, diameter 52mm, height 102mm, color of screw cap red, shapolyethyl ene assembled, CE, IVD, clinical and industrial, sterile. 1 * 264 items
Catalog # 216-2601
Supplier:  VWR Collection
Related Information