Order Entry
Finland
Orders LinkContactUsLinkComponent
Polysorbate 40

Polysorbate 40

Glass bottle, 500 mL
Catalog # L15030.AP
Polysorbate 40
Catalog # L15030.AP
CAS Number:  9005-66-7
Polysorbate 40
Polysorbate 40

Glass bottle, 500 mL

CAS Number:  9005-66-7
Price
154,00 €
Log in for availability
1 * 500 mL
Product Details & Documents

CAS No.: 9005-66-7 Molecular Formula: C62H122O26 Refractive Index: 1.4700 MDL No.: MFCD00165345

Synonyms:  Sorbitan monopalmitate, ethoxylated, Polyoxyethylene sorbitan monopalmitate
  • CAS Number:9005-66-7
  • Formula:C₆₂H₁₂₂O₂₆
  • MDL Number:MFCD00165345
  • Density:1.090
  • Storage Temperature:Ambient
Specifications

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    500 mL

Specifications