Order Entry
Finland
Orders LinkContactUsLinkComponent
Phloretin ≥98%

Phloretin ≥98%

Glass bottle for solids, 100 mg
Catalog # L10991.MC
Irritant
Phloretin ≥98%
Catalog # L10991.MC
CAS Number:  60-82-2
Phloretin ≥98%
Phloretin ≥98%

Glass bottle for solids, 100 mg

CAS Number:  60-82-2
Price
206,00 €
Log in for availability
1 * 100 mg
Product Details & Documents

Purity: 98% CAS No.: 60-82-2 Molecular Formula: C15H14O5 Formula Weight: 274.28 Storage Temperature: 2°C to 8°C Melting Point: ca 263° dec. Beilstein Reference: 1887240 Merck Reference: 14,7326 MDL No.: MFCD00002288

  • CAS Number:60-82-2
  • Formula:C₁₅H₁₄O₅
  • MDL Number:MFCD00002288
  • Molecular Weight:274.28 g/mol
  • Merck Index:14,7326
  • Melting Pt:∼ 263 °C (haj. lämpötila)
  • EINECS:200-488-7
Specifications

Specifications

  • Pakkaustyyppi:
    Glass bottle for solids
  • Pak.:
    100 mg

Specifications