Order Entry
Finland
Orders LinkContactUsLinkComponent
 

Replacement Erlenmeyer flask caps, Corning®

Vent cap, 48 mm, for 2 l flask
Catalog # 734-1215
Supplier:   Corning
Replacement Erlenmeyer flask caps, Corning®
Catalog # 734-1215
Supplier:  Corning
Replacement Erlenmeyer flask caps, Corning®
Replacement Erlenmeyer flask caps, Corning®

Vent cap, 48 mm, for 2 l flask

Price
178,00 €
Log in for availability
1 * 24 Items
Product Details & Documents

Made of PP.

Replacement caps for Corning® 500 ml, 2 and 3 litre Erlenmeyer flasks are available separately. They are supplied sterile and individually packaged.

 : This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Specifications
  • Description:
    Vent cap, 48 mm, for 2 l flask