Order Entry
Finland
Orders LinkContactUsLinkComponent
Sulphamide

Sulphamide

Glass bottle for solids, 10 g
Catalog # MAYBSB01649EA
Marketsource
Supplier:   Thermo Scientific
Sulphamide
Catalog # MAYBSB01649EA
Supplier:  Thermo Scientific
Marketsource
CAS Number:  7803-58-9
Sulphamide

Glass bottle for solids, 10 g

CAS Number:  7803-58-9
Price
Inquire For Price
Log in for availability
1 * 10 g
Product Details & Documents
Synonyms:  Sulphuric diamide
  • CAS Number:7803-58-9
  • Formula:H₄N₂O₂S
  • MDL Number:MFCD00011606
  • Molecular Weight:96.11 g/mol
  • EINECS:232-262-9
Specifications

Specifications

  • Pack type:
    Glass bottle for solids
  • Pk:
    10 g

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.