3'-Allyl-4'-hydroxyacetophenone
Glass bottle for solids, 10 g
Catalog # MAYBKM05715EA
Marketsource
Supplier: Thermo Scientific
3'-Allyl-4'-hydroxyacetophenone
Catalog # MAYBKM05715EA
Supplier: Thermo Scientific
Marketsource
CAS Number: 1132-05-4
Jump to:
Product Details & Documents
Synonyms: 3′-Allyl-4′-hydroxyacetophenone, 1-(3-Allyl-4-Hydroxy-Phenyl)Ethanone, 1-(3-Allyl-4-Hydroxyphenyl)Ethanone, 1-[4-Hydroxy-3-(2-Propen-1-Yl)Phenyl]-Ethanone, 1-(4-Hydroxy-3-Prop-2-Enyl-Phenyl)Ethanone
- CAS Number:1132-05-4
- Formula:C₁₁H₁₂O₂
- MDL Number:MFCD00068015
Specifications
Specifications
- Pack type:Glass bottle for solids
- Pk:10 g



