Order Entry
Finland
Orders LinkContactUsLinkComponent
VWR®, Speciality pH Electrodes

VWR®, Speciality pH Electrodes

pH only half cell, 0...+100 °C, Epoxy, —, 12×150 mm, BNC, Water, creams, beverage
Catalog # 662-1804
Supplier:   VWR Collection
VWR®, Speciality pH Electrodes
Catalog # 662-1804
Supplier:  VWR Collection
VWR®, Speciality pH Electrodes
VWR®, Speciality pH Electrodes

pH only half cell, 0...+100 °C, Epoxy, —, 12×150 mm, BNC, Water, creams, beverage

Price
87,00 €
Log in for availability
1 * 1 Items
Specifications
  • Connection:
    BNC
  • Ø×L:
    12×150 mm
  • For:
    Water, creams, beverage
  • Type:
    pH only half cell
  • Material:
    Epoxy
  • Seq:
    05
  • Electrolyte:
  • Temp.:
    0...+100 °C