Replacement screw caps
GL 68, 1000 ml
Catalog # RIXI8-1000-68
Supplier: BERLIN PACKAGING RIXIUS GMBH
Jump to:
Product Details & Documents
PP, black.
- Very good chemical and thermal resistance
- Suitable for bottles 215-1821 to 215-1836 and culture bottle 215-1861 to 215-1866
- LUP lining for better sealing
Specifications
- Thread:GL 68
- Seq:06
- For bottles:1000 ml