Order Entry
Finland
Orders LinkContactUsLinkComponent
BOD bottles
BOD bottles
BOD bottles, with glass stopper, 300 ml
  HACH62106
 :   Hach
BOD bottles
  HACH62106
 :  Hach
BOD bottles
BOD bottles
BOD bottles, with glass stopper, 300 ml
233,00 €
1 * 6 Items
These are standard bottles, made of borosilicate glass, for BOD analaysis.
  • Flared lip and ground-glass stopper to prevent air entrapment and for water seal
  • Serialised bottles have silk-screened, sequential numbers and include a bar code
  • Frosted marking area
Delivery information: Stoppers are included.
  • Capacity:
    300 ml
  • Description:
    BOD bottles, with glass stopper
  • Seq:
    02