Urea ≥99.5%, Proteomics Grade
Plastic bottle for solids, 1 kg
Supplier: VWR Chemicals
Jump to:
Product Details & Documents
Commonly used to denature nucleic acids for electrophoresis and to study the secondary and tertiary structure of proteins. Effective for maintaining DNA in a highly denatured state during sequencing protocols.
Synonyms: Carbonyl diamide, Carbamide, Diaminomethanone, Carbonyldiamide
- CAS Number:57-13-6
- Formula:NH₂CONH₂
- MDL Number:MFCD00008022
- Molecular Weight:60.06 g/mol
- Merck Index:13,09935
- Boiling Pt:197 °C (1013 hPa)
- Melting Pt:132.7 °C
- Density:1.34 g/cm³ (20 °C)
- Storage Temperature:Ambient
- EINECS:200-315-5
- REACH:01-2119463277-33
Documents
Specifications
Specification Test Results
- Abs.@280 nm (5 M, Water):<0.05
- Biuret Test:Pass
- Chloride:≤0.0005%
- Cyanate:≤0.002%
- DNase:none detected
- Heavy Metals (as Pb):≤0.0005%
- Insolubles:<0.005%
- Iron:≤0.0001%
- Melting Point:132 – 135 °C
- Protease:none
- Purity:≥99.5%
- RNase:none detected
- Solubility (5 M, Water):Pass
Specifications
- Pack type:Plastic bottle for solids
- Pk:1 kg



