G418 disulphate
1 KIT
Catalog # EURLECM0015Z
Supplier: EUROCLONE
Jump to:
Product Details & Documents
Synonyms: G-418 sulphate, Geneticin®, G-418 disulphate, G418 sulphate
- CAS Number:108321-42-2
- Formula:C₂₀H₄₄N₄O₁₈S₂
- MDL Number:MFCD00058314
- Molecular Weight:692.72 g/mol
- Storage Temperature:Ambient
Specifications
Specifications
- Pk:1 KIT



