Order Entry
Finland
Orders LinkContactUsLinkComponent
ISE standards

ISE standards

Potassium standard solution, 100 mg/l, 100 mg/l as K, 500 mL
Catalog # HACH2351749
Supplier:   Hach
ISE standards
Catalog # HACH2351749
Supplier:  Hach
CAS Number:  
ISE standards
ISE standards

Potassium standard solution, 100 mg/l, 100 mg/l as K, 500 mL

CAS Number:  
Price
92,80 €
Log in for availability
1 * 500 mL
Product Details & Documents

ISE standards are used to calibrate ion selective electrode measurement systems by coresponding the electrode output with the concentration of the analyte. Pairs of standards with concentrations that differ by a factor of ten are used for analysis.

Specifications

Specifications

  • Concentration:
    100 mg/l as K
  • Pk:
    500 mL
  • Description:
    Potassium standard solution, 100 mg/l

Specifications

Accessories

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Sodium standard solution, 100 mg/l
    Sodium standard solution, 100 mg/l
    HACH2318149
    100 mg/l as Na
    500 ml
    Log in for availability
    1 * 500 mL - 69,00 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrogen-nitrate standard solution, 10 mg/l
    Nitrogen-nitrate standard solution, 10 mg/l
    HACH30749
    10 mg/l as NO₃-N
    500 mL
    Log in for availability
    1 * 500 mL - 46,70 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Chloride standard solution, 100 mg/l
    Chloride standard solution, 100 mg/l
    HACH2370853
    100 mg/l as Cl
    1 L
    Log in for availability
    1 * 1 L - 72,40 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrogen-nitrate standard solution, 1000 mg/l
    Nitrogen-nitrate standard solution, 1000 mg/l
    HACH1279249
    1000 mg/l as NO₃-N
    500 mL
    Log in for availability
    1 * 500 mL - 53,30 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Calcium standard solution, 10 mg/l
    Calcium standard solution, 10 mg/l
    HACH2305442
    10 mg/l as Ca
    100 mL
    Log in for availability
    1 * 100 mL - 46,60 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrogen-ammonia standard solution, 1000 mg/l
    Nitrogen-ammonia standard solution, 1000 mg/l
    HACH2354153
    1000 mg/l as NH₃-N
    1 L
    Log in for availability
    1 * 1 L - 47,60 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Potassium standard solution, 100 mg/l
    Potassium standard solution, 100 mg/l
    HACH2351749
    100 mg/l as K
    500 mL
    Log in for availability
    1 * 500 mL - 92,80 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Fluoride standard solution, 100 mg/l
    Fluoride standard solution, 100 mg/l
    HACH23249
    100 mg/l as F
    500 mL
    Log in for availability
    1 * 500 mL - 68,10 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Nitrogen-ammonia standard solution, 100 mg/l
    Nitrogen-ammonia standard solution, 100 mg/l
    HACH2406549
    100 mg/l as NH₃-N
    500 mL
    Log in for availability
    1 * 500 mL - 46,70 €

Product Information

  • Catalog #
    Concentration
    Pk
    Availability
    Price
  • Sodium standard solution, 1000 mg/l (500 ml)
    Sodium standard solution, 1000 mg/l (500 ml)
    HACH1474949
    1000 mg/l as Na
    1
    Log in for availability
    1 * 1 Items - 70,80 €

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.