Order Entry
Finland
Orders LinkContactUsLinkComponent
Cork borers
Cork borers
Cork borers (6), 5 − 11 mm
Catalog # 217-0704
Supplier:   BOCHEM
Cork borers
Catalog # 217-0704
Supplier:  BOCHEM
Cork borers
Cork borers
Cork borers (6), 5 − 11 mm
Price
175,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
Cork borers made of hard coated nickel-plated brass.
Delivery information: Include a cleaning needle.
Specifications
  • Ø:
    5 − 11 mm
  • Description:
    Cork borers (6)