Order Entry
Finland
Orders LinkContactUsLinkComponent
Copper standard solution

Copper standard solution

Copper standard solution (NIST), 10 mg/l as Cu, 100 mL
Catalog # 705-2001
Copper standard solution
Catalog # 705-2001
Supplier:  
CAS Number:  
Copper standard solution

Copper standard solution (NIST), 10 mg/l as Cu, 100 mL

CAS Number:  
Discontinued
Product Details & Documents

Standard solution for accuracy checks in copper determinations.

Specifications

Specifications

  • Concentration:
    10 mg/l as Cu
  • Pk:
    100 mL
  • Description:
    Copper standard solution (NIST)

Specifications