Order Entry
Finland
Orders LinkContactUsLinkComponent
Comparator kits for water testing
Comparator kits for water testing
Manganese Test Kit, Model MN-5, 0 - 3,0 mg/l, 100, 1 KIT
Catalog # HACH146700
Supplier:   Hach
Comparator kits for water testing
Catalog # HACH146700
Supplier:  Hach
CAS Number:  
Comparator kits for water testing
Comparator kits for water testing
Manganese Test Kit, Model MN-5, 0 - 3,0 mg/l, 100, 1 KIT
CAS Number:  
Price
333,00 €
Log in for availability
1 * 1 Each
Product Details & Documents
Qualitative results using traditional colour comparator disc methods.
  • Portable - ideal for field testing
  • Simple fast method
  • Kits contain all required reagents and apparatus in a rugged carrying case
Please note some of the reagents supplied in the kits are hazardous, and appropriate precautions should be taken for use and disposal. Please contact Avantor for details.
Specifications
Specifications
  • Pk:
    1 KIT
  • Description:
    Manganese Test Kit, Model MN-5
  • Tests per kit:
    100
  • Range:
    0 - 3,0 mg/l
Specifications
Accessories
Product Information
  • Catalog #
    Pk
    Availability
    Price
  • Ammonium molybdate reagent for phosphate determinations, 100 ml
    Ammonium molybdate reagent for phosphate determinations, 100 ml
    HACH11032H
    1
    Log in for availability
    1 * 1 Items - 29,10 €