Order Entry
Finland
Orders LinkContactUsLinkComponent
BOD bottles
BOD bottles
BOD bottle, with glass stopper, 300 ml
Catalog # HACH62100
Supplier:   Hach
BOD bottles
Catalog # HACH62100
Supplier:  Hach
BOD bottles
BOD bottles
BOD bottle, with glass stopper, 300 ml
Price
40,70 €
Log in for availability
1 * 1 Items
Product Details & Documents
These are standard bottles, made of borosilicate glass, for BOD analaysis.
  • Flared lip and ground-glass stopper to prevent air entrapment and for water seal
  • Serialised bottles have silk-screened, sequential numbers and include a bar code
  • Frosted marking area
Delivery information: Stoppers are included.
Specifications
  • Capacity:
    300 ml
  • Description:
    BOD bottle, with glass stopper
  • Seq:
    01