Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
bath Heating with relay MU 85/R diam 187mm 800 Watt 1 * 1 items
Catalog # 453-2221
Supplier:
GESTIGKEIT HARRY
Expand image
bath Heating with relay MU 85/R diam 187mm 800 Watt 1 * 1 items
Catalog # 453-2221
Supplier:
GESTIGKEIT HARRY