Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Finland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Dish, evaporating, material quartzglass, with spout, capacity 170ml, U-diameter 95mm, height 55mm, as per DIN 12336. 1 * 1 items
Catalog # 216-1285
Supplier:
QUARTZTECHNISCHE WERKSTATTEN
Expand image
Dish, evaporating, material quartzglass, with spout, capacity 170ml, U-diameter 95mm, height 55mm, as per DIN 12336. 1 * 1 items
Catalog # 216-1285
Supplier:
QUARTZTECHNISCHE WERKSTATTEN